Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-9,9-dimethyl-9H-fluorene 98%

Supplier: AMBEED, INC
Warning

A144841-10G A144841-1G A144841-250MG A144841-25G A144841-5G
76811-922EA 80 USD InStock
76811-922 76811-924 76811-926 76811-928 76811-930
4-Bromo-9,9-dimethyl-9H-fluorene 98%
4-Bromo-9,9-dimethyl-9H-fluorene
Precaución: for Research Use Only


Formula: C₁₅H₁₃Br
MDL Number: MFCD23099415
CAS Number: 942615-32-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty