Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-(trifluoromethyl)benzyl alcohol 97%

Supplier: AMBEED, INC
Warning

A145391-10G A145391-1G A145391-250MG A145391-25G
76811-952EA 41 USD InStock
76811-952 76811-954 76811-956 76811-958
4-Chloro-3-(trifluoromethyl)benzyl alcohol 97%
4-Chloro-3-(trifluoromethyl)benzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₆ClF₃O
MW: 210.5809 g/mol
MDL Number: MFCD01631538
CAS Number: 65735-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty