Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-N-(4-methoxyphenyl)-N-phenylaniline 95%

Supplier: AMBEED, INC
Warning

A148466-1G A148466-250MG A148466-5G
76812-096EA 9 USD InStock
76812-096 76812-098 76812-100
4-Methoxy-N-(4-methoxyphenyl)-N-phenylaniline 95%
4-Methoxy-N-(4-methoxyphenyl)-N-phenylaniline
Precaución: for Research Use Only


Formula: C₂₀H₁₉NO₂
MDL Number: MFCD16659084
CAS Number: 20440-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty