Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-1,7-naphthyridine 98%

Supplier: AMBEED, INC
Warning

A149995-100MG A149995-250MG A149995-1G
76812-152EA 68 USD InStock
76812-152 76812-154 77671-110
2-Chloro-1,7-naphthyridine 98%
2-Chloro-1,7-naphthyridine
Precaución: for Research Use Only


Formula: C₈H₅ClN₂
MW: 164.59 g/mol
MDL Number: MFCD11227071
CAS Number: 35192-05-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty