Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-3-(2-nitrovinyl)benzene 95%

Supplier: AMBEED, INC
Warning

A150841-1G A150841-250MG A150841-5G
76812-184EA 20 USD InStock
76812-184 76812-186 76812-188
1-Fluoro-3-(2-nitrovinyl)benzene 95%
1-Fluoro-3-(2-nitrovinyl)benzene
Precaución: for Research Use Only


Formula: C₈H₆FNO₂
MW: 167.137 g/mol
MDL Number: MFCD00176732
CAS Number: 705-84-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty