Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Cyano-4-biphenylyl-trans-4-ethylcyclohexanecarboxylate 97%

Supplier: AMBEED, INC
Warning

A152712-10G A152712-25G A152712-5G
76812-290EA 30 USD InStock
76812-290 76812-292 76812-294
4'-Cyano-4-biphenylyl-trans-4-ethylcyclohexanecarboxylate 97%
4'-Cyano-4-biphenylyl-trans-4-ethylcyclohexanecarboxylate
Precaución: for Research Use Only


Formula: C₂₂H₂₃NO₂
MDL Number: MFCD09751067
CAS Number: 67284-56-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty