Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine-2,3-dicarboxylic anhydride 95%

Supplier: AMBEED, INC
Warning

A153241-100G A153241-10G A153241-25G A153241-500G A153241-5G
76812-336EA 50 USD InStock
76812-336 76812-338 76812-340 76812-342 76812-344
Pyridine-2,3-dicarboxylic anhydride 95%
Pyridine-2,3-dicarboxylic anhydride
Precaución: for Research Use Only


Formula: C₇H₃NO₃
MW: 149.11 g/mol
Boiling Pt: 360
Melting Pt: 146
MDL Number: MFCD00005915
CAS Number: 699-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty