Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1-methyl-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: AMBEED, INC
Warning

A155541-100MG A155541-1G A155541-250MG A155541-5G
76812-444EA 10 USD InStock
76812-444 76812-446 76812-448 76812-450
5-Bromo-1-methyl-1H-pyrrolo[2,3-b]pyridine 97%
5-Bromo-1-methyl-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only


Formula: C₈H₇BrN₂
MW: 211.06 g/mol
MDL Number: MFCD14706089
CAS Number: 183208-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty