Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Amino-4-methylphenyl)ethanone 98%

Supplier: AMBEED, INC
Warning

A157838-100MG A157838-1G A157838-250MG A157838-5G
76812-546EA 25 USD InStock
76812-546 76812-548 76812-550 76812-552
1-(2-Amino-4-methylphenyl)ethanone 98%
1-(2-Amino-4-methylphenyl)ethanone
Precaución: for Research Use Only


Formula: C₉H₁₁NO
MDL Number: MFCD19204667
CAS Number: 122710-21-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty