Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)-3-fluorophenol 98%

Supplier: AMBEED, INC
Warning

A158691-1G A158691-250MG
76812-604EA 57 USD InStock
76812-604 76812-606
4-(Benzyloxy)-3-fluorophenol 98%
4-(Benzyloxy)-3-fluorophenol
Precaución: for Research Use Only


Formula: C₁₃H₁₁FO₂
MW: 218.22 g/mol
MDL Number: MFCD01814875
CAS Number: 81228-25-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty