Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-hydroxy-3-methylbutanoate 98%

Supplier: AMBEED, INC
Warning

A159342-100MG A159342-250MG
76812-642EA 37 USD InStock
76812-642 76812-644
Ethyl 3-hydroxy-3-methylbutanoate 98%
Ethyl 3-hydroxy-3-methylbutanoate
Precaución: For research use only.


Formula: C₇H₁₄O₃
MW: 146.18 g/mol
MDL Number: MFCD16749012
CAS Number: 18267-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty