Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-iodo-1-(trifluoromethyl)benzene 95%

Supplier: AMBEED, INC
Warning

A161842-1G A161842-25G A161842-5G
76812-794EA 26 USD InStock
76812-794 76812-796 76812-798
4-Bromo-2-iodo-1-(trifluoromethyl)benzene 95%
4-Bromo-2-iodo-1-(trifluoromethyl)benzene
Precaución: for Research Use Only


Formula: C₇H₃BrF₃I
MDL Number: MFCD21603817
CAS Number: 1256945-00-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty