Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-fluoronicotinonitrile 95%

Supplier: AMBEED, INC
Warning

A162319-100G A162319-100MG A162319-10G A162319-1G A162319-250MG A162319-25G A162319-5G
76812-812EA 924 USD InStock
76812-812 76812-814 76812-816 76812-818 76812-820 76812-822 76812-824
2-Chloro-5-fluoronicotinonitrile 95%
2-Chloro-5-fluoronicotinonitrile
Precaución: for Research Use Only


Formula: C₆H₂ClFN₂
MW: 156.55 g/mol
MDL Number: MFCD15526908
CAS Number: 791644-48-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty