Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(4-hydroxyphenoxy)propanoate 95%

Supplier: AMBEED, INC
Warning

A162510-100G A162510-10G A162510-25G A162510-5G
76812-830EA 57 USD InStock
76812-830 76812-832 76812-834 76812-836
Ethyl 2-(4-hydroxyphenoxy)propanoate 95%
Ethyl 2-(4-hydroxyphenoxy)propanoate
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₄
MDL Number: MFCD23135279
CAS Number: 65343-67-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty