Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-oxotetrahydrofuran-3-carboxylate 98%

Supplier: AMBEED, INC
Warning

A163198-1G A163198-250MG A163198-5G
76812-860EA 110 USD InStock
76812-860 76812-862 77671-302
Ethyl 2-oxotetrahydrofuran-3-carboxylate 98%
Ethyl 2-oxotetrahydrofuran-3-carboxylate
Precaución: for Research Use Only


Formula: C₇H₁₀O₄
MDL Number: MFCD01725680
CAS Number: 77513-58-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty