Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Asp(OMe)-OH 97%

Supplier: AMBEED, INC
Warning

A163696-100G A163696-10G A163696-1G A163696-25G A163696-500G A163696-5G
76812-890EA 107 USD InStock
76812-890 76812-892 76812-894 76812-896 76812-898 76812-900
Boc-Asp(OMe)-OH 97%
Boc-Asp(OMe)-OH
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₆
MDL Number: MFCD00078971
CAS Number: 59768-74-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty