Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-methylbenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A164854-10G A164854-1G A164854-250MG A164854-25G
76812-946EA 43 USD InStock
76812-946 76812-948 76812-950 76812-952
4-Bromo-2-methylbenzyl alcohol 98%
4-Bromo-2-methylbenzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₉BrO
MDL Number: MFCD11847398
CAS Number: 17100-58-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty