Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Panobinostat 98%

Supplier: AMBEED, INC
Danger

A165763-10MG A165763-1MG A165763-250MG A165763-25MG A165763-5G
76813-000EA 25 USD InStock
76813-000 76813-002 76813-004 76813-006 77671-328
Panobinostat 98%
Panobinostat
Precaución: For research use only.


Formula: C₂₁H₂₃N₃O₂
MW: 349.43 g/mol
MDL Number: MFCD09833242
CAS Number: 404950-80-7
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty