Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Diphenylphosphino-2'-methylbiphenyl 97%

Supplier: AMBEED, INC
Warning

A168072-10G A168072-1G A168072-250MG A168072-25G A168072-5G
76813-106EA 57 USD InStock
76813-106 76813-108 76813-110 76813-112 76813-114
2-Diphenylphosphino-2'-methylbiphenyl 97%
2-Diphenylphosphino-2'-methylbiphenyl
Precaución: for Research Use Only


Formula: C₂₅H₂₁P
MDL Number: MFCD19441232
CAS Number: 402822-72-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty