Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromophenyl)-2-oxoacetic acid 95%

Supplier: AMBEED, INC
Warning

A173687-100MG A173687-1G A173687-250MG
76813-304EA 22 USD InStock
76813-304 76813-306 76813-308
2-(4-Bromophenyl)-2-oxoacetic acid 95%
2-(4-Bromophenyl)-2-oxoacetic acid
Precaución: for Research Use Only


Formula: C₈H₅BrO₃
MDL Number: MFCD07698695
CAS Number: 7099-87-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty