Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Methylamino)-1-(methylthio)-2-nitroethene 97%

Supplier: AMBEED, INC
Danger

A174189-1000G A174189-100G A174189-25G A174189-500G A174189-5G
76813-316EA 139 USD InStock
76813-316 76813-318 76813-320 76813-322 76813-324
1-(Methylamino)-1-(methylthio)-2-nitroethene 97%
1-(Methylamino)-1-(methylthio)-2-nitroethene
Precaución: for Research Use Only


Formula: C₄H₈N₂O₂S
MDL Number: MFCD00040994
CAS Number: 61832-41-5
UN: 3335
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty