Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-DL-alanine 97%

Supplier: AMBEED, INC
Warning

A174937-100G A174937-10G A174937-25G A174937-5G
76813-334EA 45 USD InStock
76813-334 76813-336 76813-338 76813-340
N-Methyl-DL-alanine 97%
N-Methyl-DL-alanine
Precaución: for Research Use Only


Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00063136
CAS Number: 600-21-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty