Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromo-3,5-dimethylpyridine 97%

Supplier: AMBEED, INC
Warning

A175036-250MG A175036-5G A175036-10G
76813-346EA 7 USD InStock
76813-346 76813-348 77567-584
2,6-Dibromo-3,5-dimethylpyridine 97%
2,6-Dibromo-3,5-dimethylpyridine
Precaución: For research use only.


Formula: C₇H₇Br₂N
MW: 264.945 g/mol
MDL Number: MFCD00234283
CAS Number: 117846-58-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty