Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine 98%

Supplier: AMBEED, INC
Warning

A175234-1G A175234-250MG A175234-5G
76813-350EA 66 USD InStock
76813-350 76813-352 77671-480
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine 98%
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine
Precaución: for Research Use Only


Formula: C₉H₁₈N₂
MDL Number: MFCD00013441
CAS Number: 51207-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty