Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-2-Amino-4-bromobutyric acid hydrobromide 90%

Supplier: AMBEED, INC
Warning

A175381-100MG A175381-1G A175381-250MG A175381-5G
76813-356EA 23 USD InStock
76813-356 76813-358 76813-360 76813-362
(S)-(+)-2-Amino-4-bromobutyric acid hydrobromide 90%
(S)-(+)-2-Amino-4-bromobutyric acid hydrobromide
Precaución: for Research Use Only


Formula: C₄H₈BrNO₂
MDL Number: MFCD01631290
CAS Number: 15159-65-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty