Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine 97%

Supplier: AMBEED, INC
Warning

A180342-100MG A180342-1G A180342-250MG A180342-5G
76813-526EA 41 USD InStock
76813-526 76813-528 76813-530 76813-532
5-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine 97%
5-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
Precaución: for Research Use Only


Formula: C₇H₆BrN₃
MDL Number: MFCD16657201
CAS Number: 219762-28-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty