Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-1-chloro-4-isopropylbenzene 95%

Supplier: AMBEED, INC
Warning

A181841-100MG A181841-1G A181841-250MG A181841-5G
76813-598EA 23 USD InStock
76813-598 76813-600 76813-602 76813-604
2-Bromo-1-chloro-4-isopropylbenzene 95%
2-Bromo-1-chloro-4-isopropylbenzene
Precaución: for Research Use Only


Formula: C₉H₁₀BrCl
MDL Number: MFCD11845950
CAS Number: 90350-25-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty