Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dihydroxybenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A181996-100MG A181996-10G A181996-1G A181996-250MG A181996-25G A181996-5G
76813-608EA 5 USD InStock
76813-608 76813-610 76813-612 76813-614 76813-616 76813-618
2,6-Dihydroxybenzaldehyde 98%
2,6-Dihydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₆O₃
MDL Number: MFCD08061915
CAS Number: 387-46-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty