Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromoquinoline-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A183735-100MG A183735-10G A183735-1G A183735-250MG A183735-25G A183735-5G
76813-646EA 10 USD InStock
76813-646 76813-648 76813-650 76813-652 76813-654 76813-656
7-Bromoquinoline-2-carboxylic acid 97%
7-Bromoquinoline-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₆BrNO₂
MW: 252.07 g/mol
MDL Number: MFCD11108693
CAS Number: 1057217-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty