Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-5-iodo-2,6-dimethylpyrimidine 97%

Supplier: AMBEED, INC
Warning

A183784-100MG A183784-1G
76813-664EA 18 USD InStock
76813-664 76813-666
4-Chloro-5-iodo-2,6-dimethylpyrimidine 97%
4-Chloro-5-iodo-2,6-dimethylpyrimidine
Precaución: for Research Use Only


Formula: C₆H₆ClIN₂
MW: 268.48 g/mol
MDL Number: MFCD02081997
CAS Number: 83410-16-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty