Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylphenylboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A184070-100G A184070-1G A184070-25G A184070-5G
76813-678EA 115 USD InStock
76813-678 76813-680 76813-682 76813-684
2-Methylphenylboronic acid pinacol ester 97%
2-Methylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₉BO₂
MW: 218.1
MDL Number: MFCD06795658
CAS Number: 195062-59-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty