Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-bromo-2-chloropyridine 98%

Supplier: AMBEED, INC
Danger

A187446-100G A187446-10G A187446-1G A187446-25G A187446-5G
76813-794EA 83 USD InStock
76813-794 76813-796 76813-798 76813-800 76813-802
3-Amino-5-bromo-2-chloropyridine 98%
3-Amino-5-bromo-2-chloropyridine
Precaución: for Research Use Only


Formula: C₅H₄BrClN₂
MW: 207.46
MDL Number: MFCD02682092
CAS Number: 588729-99-1
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty