Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitroisophthalic acid 98%

Supplier: AMBEED, INC
Warning

A191344-1000G A191344-100G A191344-500G
76813-898EA 31 USD InStock
76813-898 76813-900 76813-902 76813-904
5-Nitroisophthalic acid 98%
5-Nitroisophthalic acid
Precaución: for Research Use Only


Formula: C₈H₅NO₆
MDL Number: MFCD00007254
CAS Number: 618-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty