Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4-Hydroxyphenyl)valeric acid 98%

Supplier: AMBEED, INC
Warning

A191897-1G A191897-250MG A191897-5G
76813-924EA 33 USD InStock
76813-924 76813-926 76813-928
5-(4-Hydroxyphenyl)valeric acid 98%
5-(4-Hydroxyphenyl)valeric acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₃
MDL Number: MFCD04039777
CAS Number: 4654-08-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty