Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Potassium 2-(4-hydroxyphenyl)-2-((4-methoxy-4-oxobut-2-en-2-yl)amino)acetate 97%

Supplier: AMBEED, INC
Warning

A192314-100G A192314-25G A192314-500G
76813-950EA 52 USD InStock
76813-950 76813-952 76813-954 76813-956 76813-958
(R)-Potassium 2-(4-hydroxyphenyl)-2-((4-methoxy-4-oxobut-2-en-2-yl)amino)acetate 97%
(R)-Potassium 2-(4-hydroxyphenyl)-2-((4-methoxy-4-oxobut-2-en-2-yl)amino)acetate
Precaución: for Research Use Only


Formula: C₁₃H₁₄KNO₅
MDL Number: MFCD00038571
CAS Number: 69416-61-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty