Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A195174-100MG A195174-1G A195174-250MG
76814-048EA 28 USD InStock
76814-048 76814-050 76814-052
1-(2-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride 95%
1-(2-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₀F₃N·ClH
MDL Number: MFCD09028131
CAS Number: 1228879-24-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty