Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

COMU 98%

Supplier: AMBEED, INC
Danger

A195774-100G A195774-1G A195774-25G A195774-500G A195774-5G
76814-066EA 83 USD InStock
76814-066 76814-068 76814-070 76814-072 76814-074
COMU 98%
COMU
Precaución: for Research Use Only


Formula: C₁₂H₁₉F₆N₄O₄P
MW: 428.272 g/mol
MDL Number: MFCD11975052
CAS Number: 1075198-30-9
UN: 1325
ADR: 4.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty