Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-4-Amino-3-hydroxybutanoic acid 95%

Supplier: AMBEED, INC
Warning

A210658-10G A210658-1G A210658-250MG A210658-25G A210658-5G
76814-530EA 307 USD InStock
76814-530 76814-532 76814-534 76814-536 76814-538
(S)-4-Amino-3-hydroxybutanoic acid 95%
(S)-4-Amino-3-hydroxybutanoic acid
Precaución: for Research Use Only


Formula: C₄H₉NO₃
MDL Number: MFCD00672884
CAS Number: 7013-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty