Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,2S)-trans-2-Aminocyclopentanol hydrochloride 98%

Supplier: AMBEED, INC
Warning

A212536-10G A212536-1G A212536-250MG A212536-25G A212536-5G
76814-614EA 60 USD InStock
76814-614 76814-616 76814-618 76814-620 76814-622
(1S,2S)-trans-2-Aminocyclopentanol hydrochloride 98%
(1S,2S)-trans-2-Aminocyclopentanol hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂ClNO
MDL Number: MFCD07370092
CAS Number: 68327-04-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty