Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-((tert-Butyldimethylsilyl)oxy)phenyl)boronic acid 98%

Supplier: AMBEED, INC
Warning

A212714-100MG A212714-1G A212714-250MG
76814-632EA 15 USD InStock
76814-632 76814-634 76814-636
(2-((tert-Butyldimethylsilyl)oxy)phenyl)boronic acid 98%
(2-((tert-Butyldimethylsilyl)oxy)phenyl)boronic acid
Precaución: for Research Use Only


Formula: C₁₂H₂₁BO₃Si
MDL Number: MFCD18783118
CAS Number: 929277-63-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty