Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2,3-dichlorophenol 97%

Supplier: AMBEED, INC
Warning

A214395-100MG A214395-10G A214395-1G A214395-250MG A214395-5G
76814-662EA 22 USD InStock
76814-662 76814-664 76814-666 76814-668 76814-670
4-Amino-2,3-dichlorophenol 97%
4-Amino-2,3-dichlorophenol
Precaución: for Research Use Only


Formula: C₆H₅Cl₂NO
MDL Number: MFCD14455913
CAS Number: 39183-17-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty