Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(cis-2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl)methyl benzoate 95%

Supplier: AMBEED, INC
Warning

A215519-25G A215519-5G A215519-100G
76814-722EA 103 USD InStock
76814-722 76814-724 77671-930
(cis-2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl)methyl benzoate 95%
(cis-2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl)methyl benzoate
Precaución: for Research Use Only


Formula: C₁₈H₁₅BrCl₂O₄
MDL Number: MFCD07370171
CAS Number: 61397-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty