Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Pyridin-2-yl)cyclopropanecarboxylic acid 96%

Supplier: AMBEED, INC
Warning

A220760-100MG A220760-1G A220760-250MG A220760-5G
76814-864EA 20 USD InStock
76814-864 76814-866 76814-868 76814-870
1-(Pyridin-2-yl)cyclopropanecarboxylic acid 96%
1-(Pyridin-2-yl)cyclopropanecarboxylic acid
Precaución: for Research Use Only


Formula: C₉H₉NO₂
MW: 163.1733 g/mol
MDL Number: MFCD09864786
CAS Number: 162960-26-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty