Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2-Hydroxy-3-phenylpropionic acid 97%

Supplier: AMBEED, INC
Warning

A221199-100G A221199-10G A221199-1G A221199-25G A221199-5G
76814-890EA 139 USD InStock
76814-890 76814-892 76814-894 76814-896 77671-964
(R)-(+)-2-Hydroxy-3-phenylpropionic acid 97%
(R)-(+)-2-Hydroxy-3-phenylpropionic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.1739 g/mol
MDL Number: MFCD00078062
CAS Number: 7326-19-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty