Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-6-(trifluoromethyl)phenol 95%

Supplier: AMBEED, INC
Warning

A227670-100MG A227670-1G A227670-250MG A227670-5G
76815-084EA 35 USD InStock
76815-084 76815-086 76815-088 76815-090
4-Bromo-2-fluoro-6-(trifluoromethyl)phenol 95%
4-Bromo-2-fluoro-6-(trifluoromethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₃BrF₄O
MDL Number: MFCD16295240
CAS Number: 1224604-17-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty