Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethynyl-1-fluoro-4-nitrobenzene 95%

Supplier: AMBEED, INC
Warning

A232110-100MG A232110-1G A232110-250MG A232110-5G
76815-174EA 29 USD InStock
76815-174 76815-176 76815-178 76815-180
2-Ethynyl-1-fluoro-4-nitrobenzene 95%
2-Ethynyl-1-fluoro-4-nitrobenzene
Precaución: for Research Use Only


Formula: C₈H₄FNO₂
MDL Number: MFCD08703483
CAS Number: 343866-99-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty