Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyl 2,2-difluorocyclopropanecarboxylate 98%

Supplier: AMBEED, INC
Warning

A236656-250MG A236656-5G A236656-1G
76815-298EA 74 USD InStock
76815-298 76815-300 77672-110
Butyl 2,2-difluorocyclopropanecarboxylate 98%
Butyl 2,2-difluorocyclopropanecarboxylate
Precaución: for Research Use Only


Formula: C₈H₁₂F₂O₂
MDL Number: MFCD06797747
CAS Number: 260352-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty