Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-tert-Butyl 3-formylpyrrolidine-1-carboxylate 98%

Supplier: AMBEED, INC
Warning

A237097-100MG A237097-1G
76815-306EA 20 USD InStock
76815-306 76815-308
(S)-tert-Butyl 3-formylpyrrolidine-1-carboxylate 98%
(S)-tert-Butyl 3-formylpyrrolidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₃
MDL Number: MFCD14635729
CAS Number: 191348-04-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty