Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Chloroethyl)methanesulfonamide 98%

Supplier: AMBEED, INC
Warning

A239149-1G A239149-250MG A239149-5G
76815-352EA 36 USD InStock
76815-352 76815-354 76815-356
N-(2-Chloroethyl)methanesulfonamide 98%
N-(2-Chloroethyl)methanesulfonamide
Precaución: for Research Use Only


Formula: C₃H₈ClNO₂S
MDL Number: MFCD00129996
CAS Number: 21896-59-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty