Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-amino-4-methylpiperidine 97%

Supplier: AMBEED, INC
Warning

A240268-100G A240268-10G A240268-1G A240268-250MG A240268-25G A240268-5G
76815-384EA 1844 USD InStock
76815-384 76815-386 76815-388 76815-390 76815-392 76815-394
1-Boc-4-amino-4-methylpiperidine 97%
1-Boc-4-amino-4-methylpiperidine
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD03265497
CAS Number: 343788-69-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty